SY3H3D67A180 Synthonix Corporation

2-Chloro-3-fluoro-6-methylpyridine

  • MDL number MFCD03095216
  • Cas Number : 374633-32-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form White to Pale-yellow Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42573.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42573.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart