SY3H3D67A181 Synthonix Corporation

2-Chloro-5-fluoro-4-methylpyridine

  • MDL number MFCD09864695
  • Cas Number : 881891-83-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Colorless to white to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42574.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42574.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart