SY3H3D67A186 Synthonix Corporation

3-Chloro-5-iodoaniline

  • MDL number MFCD18393313
  • Cas Number : 83171-49-7
  • Molecular Weight : 253.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-chloro-5-iodoaniline
Inchi Code 1S/C6H5ClIN/c7-4-1-5(8)3-6(9)2-4/h1-3H,9H2
Inchi Key MFRQKPOHPGXPSQ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/42579.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42579.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart