SY3H3D67A191 Synthonix Corporation

(6-Chloro-3-fluoropyridin-2-yl)methanol

  • Cas Number : 884494-80-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/42671.html

Safety Information

MSDS https://synthonix.com/42671.html
Availability details for
Quantity
  • Availability
  • Add to Cart