SY3H3D67A192 Synthonix Corporation

2-Chloro-3-iodo-4-methoxypyridine

  • MDL number MFCD18415710
  • Cas Number : 1163693-01-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42673.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42673.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart