SY3H3D67A19B Synthonix Corporation

3-Chloro-5-fluorobenzene-1,2-diamine

  • Cas Number : 153505-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/42758.html

Safety Information

MSDS https://synthonix.com/42758.html
Availability details for
Quantity
  • Availability
  • Add to Cart