SY3H3D67A19E Synthonix Corporation

(2-Chloro-5-fluorophenyl)hydrazine hydrochloride

  • Cas Number : 502496-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/42761.html

Safety Information

MSDS https://synthonix.com/42761.html
Availability details for
Quantity
  • Availability
  • Add to Cart