SY3H3D67A1A1 Synthonix Corporation

(4-Chloro-2-fluorophenyl)hydrazine hydrochloride

  • MDL number MFCD06245512
  • Cas Number : 64172-78-7
  • Molecular Weight : 197.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42764.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42764.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart