SY3H3D67A1A8 Synthonix Corporation

5-Chloro-4-methyl-3-nitropyridin-2-amine

  • MDL number MFCD07778624
  • Cas Number : 148612-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Yellow solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42817.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42817.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart