SY3H3D67A1AA Synthonix Corporation

4-Chloro-6-methoxy-2-methyl-5-nitropyrimidine

  • MDL number MFCD29922356
  • Cas Number : 60331-15-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42822.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42822.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart