SY3H3D67A1F5 Synthonix Corporation

2-Chloro-4-methyl-6-(methylthio)pyrimidine

  • Cas Number : 89466-59-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43007.html

Safety Information

MSDS https://synthonix.com/43007.html
Availability details for
Quantity
  • Availability
  • Add to Cart