SY3H3D67A207 Synthonix Corporation

2-Chloro-4-(methoxymethyl)pyrimidine

  • Cas Number : 1289387-30-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/43072.html

Safety Information

MSDS https://synthonix.com/43072.html
Availability details for
Quantity
  • Availability
  • Add to Cart