SY3H3D67A208 Synthonix Corporation

2-Chloro-4-methoxy-6-methylpyrimidine

  • MDL number MFCD08689881
  • Cas Number : 22536-64-7
  • Molecular Weight : 158.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43073.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43073.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart