SY3H3D67A24A Synthonix Corporation

3-Chloro-N,N-dimethylpyrazin-2-amine

  • MDL number MFCD12827499
  • Cas Number : 54126-45-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43272.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43272.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart