SY3H3D67A265 Synthonix Corporation

1-(Cyanomethyl)pyrrolidin-1-ium trifluoromethanesulfonate

  • MDL number MFCD23726589
  • Cas Number : 573987-48-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43583.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43583.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart