SY3H3D67A267 Synthonix Corporation

4-Chloro-N-ethyl-6-methylpyrimidin-2-amine

  • MDL number MFCD01150900
  • Cas Number : 5748-33-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43692.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43692.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart