SY3H3D67A270 Synthonix Corporation

2-(2-Chloroethoxy)tetrahydro-2H-pyran

  • Cas Number : 5631-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/43866.html

Safety Information

MSDS https://synthonix.com/43866.html
Availability details for
Quantity
  • Availability
  • Add to Cart