SY3H3D67A272 Synthonix Corporation

1-(3-Chloropropyl)pyrrolidine

  • MDL number MFCD07787234
  • Cas Number : 39743-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. refrigerator
Physical Form Colorless to light-yellow to yellow liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43884.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43884.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart