SY3H3D67A27A Synthonix Corporation

6-Chloro-3-(trifluoromethyl)picolinonitrile

  • MDL number MFCD18261725
  • Cas Number : 401590-41-8
  • Molecular Weight : 206.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43967.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43967.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart