SY3H3D67A286 Synthonix Corporation

5-Chloro-2,4-difluorobenzonitrile

  • Cas Number : 146780-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/44001.html

Safety Information

MSDS https://synthonix.com/44001.html
Availability details for
Quantity
  • Availability
  • Add to Cart