SY3H3D67A28B Synthonix Corporation

2-chloro-1-fluoro-4-(trifluoromethyl)benzene

  • MDL number No MDL found
  • Cas Number : 78068-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44041.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44041.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart