SY3H3D67A290 Synthonix Corporation

2-Chloro-3-nitrobenzonitrile

  • Cas Number : 34662-24-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/44060.html

Safety Information

MSDS https://synthonix.com/44060.html
Availability details for
Quantity
  • Availability
  • Add to Cart