SY3H3D67A292 Synthonix Corporation

3-Chloro-5-nitrobenzonitrile

  • Cas Number : 34662-30-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/44062.html

Safety Information

MSDS https://synthonix.com/44062.html
Availability details for
Quantity
  • Availability
  • Add to Cart