SY3H3D67A2EB Synthonix Corporation

2-Chloro-4,5-difluorobenzaldehyde

  • MDL number MFCD07782050
  • Cas Number : 165047-23-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44262.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44262.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart