SY3H3D67A2F4 Synthonix Corporation

5-Chloro-3-iodo-1H-pyrrolo[2,3-c]pyridine

  • MDL number MFCD12963382
  • Cas Number : 1190310-51-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44335.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44335.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart