SY3H3D67A2FC Synthonix Corporation

2-chloro-4-nitro-6-(trifluoromethyl)aniline

  • Cas Number : 400-67-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/44354.html

Safety Information

MSDS https://synthonix.com/44354.html
Availability details for
Quantity
  • Availability
  • Add to Cart