SY3H3D67A2FF Synthonix Corporation

2-Cyano-3-(thiazol-2-yl)acrylic acid

  • Cas Number : 1567534-28-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/44360.html

Safety Information

MSDS https://synthonix.com/44360.html
Availability details for
Quantity
  • Availability
  • Add to Cart