SY3H3D67A30F Synthonix Corporation

3-Chloro-5-ethynylpyridine

  • Cas Number : 329202-22-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/44423.html

Safety Information

MSDS https://synthonix.com/44423.html
Availability details for
Quantity
  • Availability
  • Add to Cart