SY3H3D67A310 Synthonix Corporation

5-Chloro-2-ethynylpyridine hydrochloride

  • Cas Number : 1956341-52-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/44424.html

Safety Information

MSDS https://synthonix.com/44424.html
Availability details for
Quantity
  • Availability
  • Add to Cart