SY3H3D67A334 Synthonix Corporation

6-Chlorobenzo[d]thiazole-2-thiol

  • MDL number MFCD00665746
  • Cas Number : 51618-29-2
  • Molecular Weight : 201.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-chloro-1,3-benzothiazole-2(3H)-thione
Inchi Code 1S/C7H4ClNS2/c8-4-1-2-5-6(3-4)11-7(10)9-5/h1-3H,(H,9,10)
Inchi Key CLHLOHAQAADLRA-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Refrigerator
Physical Form Light yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/44433.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44433.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart