SY3H3D67A342 Synthonix Corporation

3-Chloro-4-nitrobenzoic acid

  • MDL number MFCD01632172
  • Cas Number : 39608-47-4
  • Molecular Weight : 201.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44503.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44503.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart