SY3H3D67A348 Synthonix Corporation

6-Chlorobenzoxazole

  • Cas Number : 227197-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/44529.html

Safety Information

MSDS https://synthonix.com/44529.html
Availability details for
Quantity
  • Availability
  • Add to Cart