SY3H3D67A34A Synthonix Corporation

3-Chloro-1,2-benzisoxazole

  • Cas Number : 16263-52-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/44531.html

Safety Information

MSDS https://synthonix.com/44531.html
Availability details for
Quantity
  • Availability
  • Add to Cart