SY3H3D67A351 Synthonix Corporation

4-Chlorobenzo[d]thiazol-2(3H)-one

  • MDL number MFCD00796306
  • Cas Number : 39205-62-4
  • Molecular Weight : 185.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44540.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/44540.html
pictograms GHS07
hstatements H302, H315, H319, H332, H335
Availability details for
Quantity
  • Availability
  • Add to Cart