SY3H3D67A352 Synthonix Corporation

7-Chlorobenzo[d]thiazol-2(3H)-one

  • Cas Number : 80416-76-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/44541.html

Safety Information

MSDS https://synthonix.com/44541.html
Availability details for
Quantity
  • Availability
  • Add to Cart