SY3H3D67A355 Synthonix Corporation

2-Chloro-3-fluorobenzoic acid

  • MDL number MFCD00973903
  • Cas Number : 102940-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to very pale yellow or pale-red powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44588.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44588.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart