SY3H3D67A35E Synthonix Corporation

3-Chloro-2-iodobenzoic acid

  • MDL number MFCD06808824
  • Cas Number : 123278-03-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44599.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44599.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart