SY3H3D67A364 Synthonix Corporation

5-(Chlorosulfonyl)-2-fluorobenzoic acid

  • MDL number MFCD03030335
  • Cas Number : 37098-75-2
  • Molecular Weight : 238.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44630.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44630.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart