SY3H3D67A37A Synthonix Corporation

3-Chloropyrazolo[1,5-a]pyridine

  • MDL number MFCD28902441
  • Cas Number : 1799439-18-1
  • Molecular Weight : 152.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44777.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44777.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart