SY3H3D67A39D Synthonix Corporation

2-Chloro-4-methylpyridine-3-carbonitrile

  • MDL number MFCD07082888
  • Cas Number : 65169-38-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Pale-yellow solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44784.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44784.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart