SY3H3D67A3A1 Synthonix Corporation

7-chloro-benzothiazol-6-ylamine

  • Cas Number : 70202-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/44790.html

Safety Information

MSDS https://synthonix.com/44790.html
Availability details for
Quantity
  • Availability
  • Add to Cart