SY3H3D67A3AB Synthonix Corporation

6-Cyanopicolinamide

  • MDL number MFCD16038715
  • Cas Number : 52368-18-0
  • Molecular Weight : 147.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-cyanopicolinamide
Inchi Code 1S/C7H5N3O/c8-4-5-2-1-3-6(10-5)7(9)11/h1-3H,(H2,9,11)
Inchi Key RIQROIGOHGXLPI-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/44865.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44865.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart