SY3H3D67A3B2 Synthonix Corporation

3-Chloro-5-trifluoromethylaniline

  • MDL number MFCD02684210
  • Cas Number : 69411-05-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form White to Pale-yellow to Yellow-brown Solid or Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44911.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44911.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart