SY3H3D67A3B5 Synthonix Corporation

2-Chloro-5-(trifluoromethyl)aniline

  • MDL number No MDL found
  • Cas Number : 121-50-6
  • Molecular Weight : 195.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44913.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44913.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart