SY3H3D67A3B7 Synthonix Corporation

3-Chloro-4-(trifluoromethyl)aniline

  • MDL number MFCD00278479
  • Cas Number : 445-13-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Liquid or Semi-solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44915.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44915.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart