SY3H3D67A3C4 Synthonix Corporation

2-Chloro-1-(6-chloropyridin-3-yl)ethanone

  • Cas Number : 136592-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/45047.html

Safety Information

MSDS https://synthonix.com/45047.html
Availability details for
Quantity
  • Availability
  • Add to Cart