SY3H3D67A3CB Synthonix Corporation

5-Chloro-2-(trifluoromethoxy)aniline

  • MDL number MFCD06410919
  • Cas Number : 326-64-7
  • Molecular Weight : 211.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-2-(trifluoromethoxy)aniline
Inchi Code 1S/C7H5ClF3NO/c8-4-1-2-6(5(12)3-4)13-7(9,10)11/h1-3H,12H2
Inchi Key KSBUTBXMRFVLND-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Refrigerator
Physical Form Yellow to Brown Liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/45055.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45055.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart