SY3H3D67A3E5 Synthonix Corporation

4-Chloro-3-fluorotoluene

  • MDL number No MDL found
  • Cas Number : 5527-94-6
  • Molecular Weight : 144.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-chloro-2-fluoro-4-methylbenzene
Inchi Code 1S/C7H6ClF/c1-5-2-3-6(8)7(9)4-5/h2-4H,1H3
Inchi Key MHXNHUSVBYUTJL-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Refrigerator
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/45177.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45177.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart