SY3H3D67A3E8 Synthonix Corporation

2-Chloro-6-iodotoluene

  • MDL number MFCD00019013
  • Cas Number : 42048-11-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Clear liquid / Colour: Pale yellow - Reddish yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45180.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45180.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart