SY3H3D67A406 Synthonix Corporation

5-Chloro-2-iodotoluene

  • MDL number No MDL found
  • Cas Number : 23399-70-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/45182.html

Safety Information

MSDS https://synthonix.com/45182.html
Availability details for
Quantity
  • Availability
  • Add to Cart